Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGAGGAGGATCACAACCCAAAGT[C/G]CTCACAGAGGGAGCAGTGCTGCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARNTL2-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARNTL2-AS1 - ARNTL2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMCO2 - single-pass membrane protein with coiled-coil domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145010.1 | Intron | NP_001138482.1 | ||||
XM_011520634.2 | Intron | XP_011518936.1 | ||||
XM_011520635.2 | Intron | XP_011518937.1 | ||||
XM_011520636.2 | Intron | XP_011518938.1 | ||||
XM_017019254.1 | Intron | XP_016874743.1 | ||||
XM_017019255.1 | Intron | XP_016874744.1 |
Set Membership: |
HapMap |