Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604126 | ||||||||||||||||||||
Literature Links: |
SVIL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SVIL - supervillin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323599.1 | 6869 | Missense Mutation | TAC,TTC | Y,F 1729 | NP_001310528.1 | |
NM_001323600.1 | 6869 | Missense Mutation | TAC,TTC | Y,F 1645 | NP_001310529.1 | |
NM_003174.3 | 6869 | Missense Mutation | TAC,TTC | Y,F 1613 | NP_003165.2 | |
NM_021738.2 | 6869 | Missense Mutation | TAC,TTC | Y,F 2039 | NP_068506.2 |
SVIL-AS1 - SVIL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |