Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATTTCACCTTAAGGGCAAGGTGA[C/T]TGCTCCTTTAAGGCTCTGAAGCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609388 | ||||||||||||||||||||
Literature Links: |
LOC101927814 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927814 - uncharacterized LOC101927814 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL9 - methyltransferase like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077180.2 | Intron | NP_001070648.1 | ||||
NM_001288659.1 | Intron | NP_001275588.1 | ||||
NM_001288660.1 | Intron | NP_001275589.1 | ||||
NM_016025.4 | Intron | NP_057109.3 |