Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603621 MIM: 615216 MIM: 609172 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXH1 - forkhead box H1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIFC2 - kinesin family member C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R16A - protein phosphatase 1 regulatory subunit 16A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005272350.2 | Intron | XP_005272407.1 | ||||
XM_011517349.2 | Intron | XP_011515651.1 | ||||
XM_017013912.1 | Intron | XP_016869401.1 | ||||
XM_017013913.1 | Intron | XP_016869402.1 | ||||
XM_017013914.1 | Intron | XP_016869403.1 | ||||
XM_017013915.1 | Intron | XP_016869404.1 |
Set Membership: |
HapMap |