Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605837 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HERC2 - HECT and RLD domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004667.5 | Intron | NP_004658.3 | ||||
XM_005268276.4 | Intron | XP_005268333.1 | ||||
XM_006720726.3 | Intron | XP_006720789.1 | ||||
XM_006720727.3 | Intron | XP_006720790.1 | ||||
XM_011522131.2 | Intron | XP_011520433.1 | ||||
XM_011522133.2 | Intron | XP_011520435.1 | ||||
XM_017022695.1 | Intron | XP_016878184.1 | ||||
XM_017022696.1 | Intron | XP_016878185.1 | ||||
XM_017022697.1 | Intron | XP_016878186.1 | ||||
XM_017022698.1 | Intron | XP_016878187.1 |
Set Membership: |
HapMap |