Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603340 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ASB14 - ankyrin repeat and SOCS box containing 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAH12 - dynein axonemal heavy chain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291661.1 | Intron | NP_001278590.1 | ||||
NM_198564.3 | Intron | NP_940966.2 | ||||
XM_011533466.1 | Intron | XP_011531768.1 | ||||
XM_011533467.1 | Intron | XP_011531769.1 | ||||
XM_011533468.1 | Intron | XP_011531770.1 | ||||
XM_011533469.1 | Intron | XP_011531771.1 | ||||
XM_011533471.2 | Intron | XP_011531773.1 | ||||
XM_011533474.1 | Intron | XP_011531776.1 | ||||
XM_017005860.1 | Intron | XP_016861349.1 | ||||
XM_017005861.1 | Intron | XP_016861350.1 | ||||
XM_017005862.1 | Intron | XP_016861351.1 | ||||
XM_017005863.1 | Intron | XP_016861352.1 |
Set Membership: |
HapMap |