Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602701 | ||||||||||||||||||||
Literature Links: |
SLMAP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLMAP - sarcolemma associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304420.2 | Intron | NP_001291349.1 | ||||
NM_001304421.2 | Intron | NP_001291350.1 | ||||
NM_001304422.2 | Intron | NP_001291351.1 | ||||
NM_001304423.2 | Intron | NP_001291352.1 | ||||
NM_001311178.1 | Intron | NP_001298107.1 | ||||
NM_001311179.1 | Intron | NP_001298108.1 | ||||
NM_007159.4 | Intron | NP_009090.2 |