Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605836 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC13B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UNC13B - unc-13 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006377.3 | Intron | NP_006368.3 | ||||
XM_011517681.2 | Intron | XP_011515983.1 | ||||
XM_011517682.2 | Intron | XP_011515984.1 | ||||
XM_011517683.2 | Intron | XP_011515985.1 | ||||
XM_011517684.2 | Intron | XP_011515986.1 | ||||
XM_011517685.2 | Intron | XP_011515987.1 | ||||
XM_011517686.2 | Intron | XP_011515988.1 | ||||
XM_017014191.1 | Intron | XP_016869680.1 | ||||
XM_017014192.1 | Intron | XP_016869681.1 |