Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609759 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRDM7 - PR domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098173.1 | Intron | NP_001091643.1 | ||||
XM_005256274.3 | Intron | XP_005256331.1 | ||||
XM_011522829.1 | Intron | XP_011521131.1 | ||||
XM_011522831.2 | Intron | XP_011521133.1 | ||||
XM_017022882.1 | Intron | XP_016878371.1 | ||||
XM_017022883.1 | Intron | XP_016878372.1 | ||||
XM_017022884.1 | Intron | XP_016878373.1 |
Set Membership: |
HapMap |