Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACATTCAGGTCTCACTGAGGGGA[A/G]ATGTGCCCAGGACACTTCCACTGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147880 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL6R PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL6R - interleukin 6 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000565.3 | Intron | NP_000556.1 | ||||
NM_001206866.1 | Intron | NP_001193795.1 | ||||
NM_181359.2 | Intron | NP_852004.1 | ||||
XM_005245139.1 | Intron | XP_005245196.1 | ||||
XM_005245140.2 | Intron | XP_005245197.1 | ||||
XM_006711298.1 | Intron | XP_006711361.1 | ||||
XM_006711299.3 | Intron | XP_006711362.1 | ||||
XM_017001199.1 | Intron | XP_016856688.1 | ||||
XM_017001200.1 | Intron | XP_016856689.1 | ||||
XM_017001201.1 | Intron | XP_016856690.1 |
LOC101928101 - uncharacterized LOC101928101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |