Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120131 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COL4A4 - collagen type IV alpha 4 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000092.4 | Intron | NP_000083.3 | ||||
XM_005246281.3 | Intron | XP_005246338.1 | ||||
XM_005246282.3 | Intron | XP_005246339.1 | ||||
XM_006712246.3 | Intron | XP_006712309.1 | ||||
XM_011510557.2 | Intron | XP_011508859.1 | ||||
XM_011510558.2 | Intron | XP_011508860.1 | ||||
XM_011510559.2 | Intron | XP_011508861.1 | ||||
XM_011510560.2 | Intron | XP_011508862.1 | ||||
XM_011510561.2 | Intron | XP_011508863.1 | ||||
XM_011510562.2 | Intron | XP_011508864.1 | ||||
XM_011510565.2 | Intron | XP_011508867.1 | ||||
XM_011510566.2 | Intron | XP_011508868.1 | ||||
XM_011510567.2 | Intron | XP_011508869.1 | ||||
XM_011510568.2 | Intron | XP_011508870.1 | ||||
XM_011510569.2 | Intron | XP_011508871.1 | ||||
XM_011510570.2 | Intron | XP_011508872.1 | ||||
XM_011510572.2 | Intron | XP_011508874.1 | ||||
XM_017003296.1 | Intron | XP_016858785.1 | ||||
XM_017003297.1 | Intron | XP_016858786.1 | ||||
XM_017003298.1 | Intron | XP_016858787.1 | ||||
XM_017003299.1 | Intron | XP_016858788.1 | ||||
XM_017003300.1 | Intron | XP_016858789.1 |
RHBDD1 - rhomboid domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167608.1 | Intron | NP_001161080.1 | ||||
NM_032276.3 | Intron | NP_115652.2 | ||||
XM_017005083.1 | Intron | XP_016860572.1 | ||||
XM_017005084.1 | Intron | XP_016860573.1 | ||||
XM_017005085.1 | Intron | XP_016860574.1 | ||||
XM_017005086.1 | Intron | XP_016860575.1 | ||||
XM_017005087.1 | Intron | XP_016860576.1 | ||||
XM_017005088.1 | Intron | XP_016860577.1 | ||||
XM_017005089.1 | Intron | XP_016860578.1 | ||||
XM_017005090.1 | Intron | XP_016860579.1 | ||||
XM_017005091.1 | Intron | XP_016860580.1 | ||||
XM_017005092.1 | Intron | XP_016860581.1 | ||||
XM_017005093.1 | Intron | XP_016860582.1 | ||||
XM_017005094.1 | Intron | XP_016860583.1 | ||||
XM_017005095.1 | Intron | XP_016860584.1 |
Set Membership: |
HapMap |