Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCACAGGATAAATAAAGACGTTA[A/T]CTTAAGAGCAGTTATGTCTTACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107985388 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107985388 - pre-mRNA-splicing factor cwc22-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017028186.1 | Intron | XP_016883675.1 |
PCIF1 - PDX1 C-terminal inhibiting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022104.3 | Intron | NP_071387.1 | ||||
XM_011528980.2 | Intron | XP_011527282.1 | ||||
XM_011528981.2 | Intron | XP_011527283.1 | ||||
XM_017028013.1 | Intron | XP_016883502.1 | ||||
XM_017028014.1 | Intron | XP_016883503.1 |