Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613945 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJC5B - DnaJ heat shock protein family (Hsp40) member C5 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033105.4 | Intron | NP_149096.2 | ||||
XM_011517618.2 | Intron | XP_011515920.1 | ||||
XM_011517619.2 | Intron | XP_011515921.1 | ||||
XM_011517620.2 | Intron | XP_011515922.1 | ||||
XM_011517621.2 | Intron | XP_011515923.1 |
Set Membership: |
HapMap |