Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF16B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF16B - kinesin family member 16B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199865.1 | Intron | NP_001186794.1 | ||||
NM_001199866.1 | Intron | NP_001186795.1 | ||||
NM_024704.4 | Intron | NP_078980.3 | ||||
XM_005260750.3 | Intron | XP_005260807.1 | ||||
XM_005260751.4 | Intron | XP_005260808.1 | ||||
XM_005260752.3 | Intron | XP_005260809.1 | ||||
XM_005260753.3 | Intron | XP_005260810.1 | ||||
XM_005260754.3 | Intron | XP_005260811.1 | ||||
XM_005260755.3 | Intron | XP_005260812.1 | ||||
XM_006723588.3 | Intron | XP_006723651.1 | ||||
XM_017027926.1 | Intron | XP_016883415.1 |
Set Membership: |
HapMap |