Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614476 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CT62 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CT62 - cancer/testis antigen 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THSD4 - thrombospondin type 1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286429.1 | Intron | NP_001273358.1 | ||||
NM_024817.2 | Intron | NP_079093.2 | ||||
XM_006720692.3 | Intron | XP_006720755.1 | ||||
XM_011522043.2 | Intron | XP_011520345.1 | ||||
XM_011522044.2 | Intron | XP_011520346.1 | ||||
XM_017022582.1 | Intron | XP_016878071.1 | ||||
XM_017022583.1 | Intron | XP_016878072.1 | ||||
XM_017022584.1 | Intron | XP_016878073.1 | ||||
XM_017022585.1 | Intron | XP_016878074.1 | ||||
XM_017022586.1 | Intron | XP_016878075.1 |
Set Membership: |
HapMap |