Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 128239 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DAG1 - dystroglycan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165928.3 | 283 | UTR 5 | NP_001159400.2 | |||
NM_001177634.2 | 283 | UTR 5 | NP_001171105.1 | |||
NM_001177635.2 | 283 | Intron | NP_001171106.1 | |||
NM_001177636.2 | 283 | UTR 5 | NP_001171107.1 | |||
NM_001177637.2 | 283 | UTR 5 | NP_001171108.1 | |||
NM_001177638.2 | 283 | UTR 5 | NP_001171109.1 | |||
NM_001177639.2 | 283 | Intron | NP_001171110.1 | |||
NM_001177640.2 | 283 | UTR 5 | NP_001171111.1 | |||
NM_001177641.2 | 283 | UTR 5 | NP_001171112.1 | |||
NM_001177642.2 | 283 | UTR 5 | NP_001171113.1 | |||
NM_001177643.2 | 283 | Intron | NP_001171114.1 | |||
NM_001177644.2 | 283 | UTR 5 | NP_001171115.1 | |||
NM_004393.5 | 283 | UTR 5 | NP_004384.4 |
Set Membership: |
HapMap |