Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600843 MIM: 604328 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
P2RX3 - purinergic receptor P2X 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002559.3 | 281 | Intron | NP_002550.2 | |||
XM_011545069.2 | 281 | UTR 5 | XP_011543371.1 | |||
XM_011545070.2 | 281 | Intron | XP_011543372.1 | |||
XM_011545071.2 | 281 | Intron | XP_011543373.1 | |||
XM_011545072.2 | 281 | Intron | XP_011543374.1 | |||
XM_011545073.1 | 281 | Intron | XP_011543375.1 |
SSRP1 - structure specific recognition protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003146.2 | 281 | Intron | NP_003137.1 | |||
XM_017018180.1 | 281 | Intron | XP_016873669.1 | |||
XM_017018181.1 | 281 | Intron | XP_016873670.1 |
Set Membership: |
HapMap |