Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTACCTCTCACAAGAGACAACAG[A/C]TAGCTGCTGTTTCACAGCGTGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM156A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM156A - family with sequence similarity 156 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242489.1 | Intron | NP_001229418.1 | ||||
NM_001242490.1 | Intron | NP_001229419.1 | ||||
NM_001242491.1 | Intron | NP_001229420.1 | ||||
NM_001242492.1 | Intron | NP_001229421.1 | ||||
NM_001242493.1 | Intron | NP_001229422.1 | ||||
NM_001242494.1 | Intron | NP_001229423.1 | ||||
NM_001242495.1 | Intron | NP_001229424.1 | ||||
NM_001242496.1 | Intron | NP_001229425.1 | ||||
NM_001242497.1 | Intron | NP_001229426.1 | ||||
NM_014138.4 | Intron | NP_054857.2 |