Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608862 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPATCH4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPATCH4 - G-patch domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAPLN2 - hyaluronan and proteoglycan link protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021817.2 | Intron | NP_068589.1 | ||||
XM_005245415.2 | Intron | XP_005245472.1 | ||||
XM_011509853.2 | Intron | XP_011508155.1 | ||||
XM_017002020.1 | Intron | XP_016857509.1 | ||||
XM_017002021.1 | Intron | XP_016857510.1 |
NAXE - NAD(P)HX epimerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |