Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGCCCTGCTGAGGCTTCCGT[G/C]TAAACTCCGCAGAGCACACAGCCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612302 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADGRA1 - adhesion G protein-coupled receptor A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083909.2 | Intron | NP_001077378.1 | ||||
NM_001291085.1 | Intron | NP_001278014.1 | ||||
XM_005252695.1 | Intron | XP_005252752.1 | ||||
XM_011540273.1 | Intron | XP_011538575.1 | ||||
XM_017016779.1 | Intron | XP_016872268.1 |
ADGRA1-AS1 - adhesion G protein-coupled receptor A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |