Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607761 MIM: 104240 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIRREL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIRREL3 - kin of IRRE like 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161707.1 | Intron | NP_001155179.1 | ||||
NM_001301097.1 | Intron | NP_001288026.1 | ||||
NM_032531.3 | Intron | NP_115920.1 | ||||
XM_011543026.2 | Intron | XP_011541328.1 | ||||
XM_011543027.2 | Intron | XP_011541329.1 | ||||
XM_011543028.2 | Intron | XP_011541330.1 | ||||
XM_011543030.2 | Intron | XP_011541332.1 | ||||
XM_011543031.2 | Intron | XP_011541333.1 | ||||
XM_011543032.2 | Intron | XP_011541334.1 | ||||
XM_011543033.1 | Intron | XP_011541335.1 | ||||
XM_017018418.1 | Intron | XP_016873907.1 | ||||
XM_017018419.1 | Intron | XP_016873908.1 | ||||
XM_017018420.1 | Intron | XP_016873909.1 |
ST3GAL4 - ST3 beta-galactoside alpha-2,3-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254757.1 | Intron | NP_001241686.1 | ||||
NM_001254758.1 | Intron | NP_001241687.1 | ||||
NM_001254759.1 | Intron | NP_001241688.1 | ||||
NM_006278.2 | Intron | NP_006269.1 | ||||
XM_011542959.1 | Intron | XP_011541261.1 | ||||
XM_011542960.2 | Intron | XP_011541262.1 | ||||
XM_017018134.1 | Intron | XP_016873623.1 |
Set Membership: |
HapMap |