Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608139 MIM: 605947 | ||||||||||||||||||||
Literature Links: |
CENPV PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPV - centromere protein V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181716.2 | 793 | Missense Mutation | CAG,CGG | Q,R 244 | NP_859067.2 |
PIGL - phosphatidylinositol glycan anchor biosynthesis class L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004278.3 | 793 | Intron | NP_004269.1 | |||
XM_011524080.2 | 793 | Intron | XP_011522382.1 | |||
XM_017025349.1 | 793 | Intron | XP_016880838.1 | |||
XM_017025350.1 | 793 | Intron | XP_016880839.1 | |||
XM_017025351.1 | 793 | Intron | XP_016880840.1 | |||
XM_017025352.1 | 793 | Intron | XP_016880841.1 | |||
XM_017025353.1 | 793 | Intron | XP_016880842.1 | |||
XM_017025354.1 | 793 | Intron | XP_016880843.1 | |||
XM_017025355.1 | 793 | Intron | XP_016880844.1 | |||
XM_017025356.1 | 793 | Intron | XP_016880845.1 |