Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180960 MIM: 600201 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCY PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AHCY - adenosylhomocysteinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000687.3 | Intron | NP_000678.1 | ||||
NM_001161766.1 | Intron | NP_001155238.1 | ||||
NM_001322084.1 | Intron | NP_001309013.1 | ||||
NM_001322085.1 | Intron | NP_001309014.1 | ||||
NM_001322086.1 | Intron | NP_001309015.1 | ||||
XM_005260317.2 | Intron | XP_005260374.1 | ||||
XM_011528656.2 | Intron | XP_011526958.2 | ||||
XM_011528657.2 | Intron | XP_011526959.2 | ||||
XM_011528658.2 | Intron | XP_011526960.2 | ||||
XM_011528659.1 | Intron | XP_011526961.1 | ||||
XM_017027708.1 | Intron | XP_016883197.1 | ||||
XM_017027709.1 | Intron | XP_016883198.1 | ||||
XM_017027710.1 | Intron | XP_016883199.1 |
ASIP - agouti signaling protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001672.2 | Intron | NP_001663.2 | ||||
XM_005260412.3 | Intron | XP_005260469.1 | ||||
XM_011528820.2 | Intron | XP_011527122.1 | ||||
XM_011528821.1 | Intron | XP_011527123.1 |
Set Membership: |
HapMap |