Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615154 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DYDC1 - DPY30 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001269053.1 | Intron | NP_001255982.1 | ||||
NM_138812.3 | Intron | NP_620167.1 | ||||
XM_005269549.4 | Intron | XP_005269606.1 | ||||
XM_005269550.4 | Intron | XP_005269607.1 | ||||
XM_011539334.2 | Intron | XP_011537636.1 | ||||
XM_011539335.1 | Intron | XP_011537637.1 | ||||
XM_017015749.1 | Intron | XP_016871238.1 |
DYDC2 - DPY30 domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270041.1 | Intron | NP_001256970.1 | ||||
NM_001270042.1 | Intron | NP_001256971.1 | ||||
NM_032372.5 | Intron | NP_115748.1 | ||||
XM_011540267.2 | Intron | XP_011538569.1 | ||||
XM_011540268.1 | Intron | XP_011538570.1 | ||||
XM_011540270.2 | Intron | XP_011538572.1 |
Set Membership: |
HapMap |