Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611749 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZRANB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZRANB1 - zinc finger RANBP2-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017580.2 | Intron | NP_060050.2 | ||||
XM_005269925.4 | Intron | XP_005269982.1 | ||||
XM_005269926.4 | Intron | XP_005269983.1 | ||||
XM_006717907.2 | Intron | XP_006717970.1 | ||||
XM_011539887.2 | Intron | XP_011538189.1 | ||||
XM_017016357.1 | Intron | XP_016871846.1 | ||||
XM_017016358.1 | Intron | XP_016871847.1 |
Set Membership: |
HapMap |