Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606598 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GDAP1 - ganglioside induced differentiation associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040875.2 | Intron | NP_001035808.1 | ||||
NM_018972.2 | Intron | NP_061845.2 | ||||
XM_011517551.2 | Intron | XP_011515853.1 | ||||
XM_011517552.2 | Intron | XP_011515854.1 | ||||
XM_017013585.1 | Intron | XP_016869074.1 | ||||
XM_017013586.1 | Intron | XP_016869075.1 |