Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616965 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRG5 - adhesion G protein-coupled receptor G5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304376.2 | Intron | NP_001291305.1 | ||||
NM_001318481.1 | Intron | NP_001305410.1 | ||||
NM_153837.3 | Intron | NP_722579.1 | ||||
XM_011522949.2 | Intron | XP_011521251.1 | ||||
XM_011522950.2 | Intron | XP_011521252.1 |
CCDC102A - coiled-coil domain containing 102A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033212.3 | Intron | NP_149989.2 | ||||
XM_011523469.2 | Intron | XP_011521771.1 |
Set Membership: |
HapMap |