Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606847 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCOF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCOF1 - treacle ribosome biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000356.3 | 20 | UTR 5 | NP_000347.2 | |||
NM_001008657.2 | 20 | UTR 5 | NP_001008657.1 | |||
NM_001135243.1 | 20 | UTR 5 | NP_001128715.1 | |||
NM_001135244.1 | 20 | UTR 5 | NP_001128716.1 | |||
NM_001135245.1 | 20 | UTR 5 | NP_001128717.1 | |||
NM_001195141.1 | 20 | UTR 5 | NP_001182070.1 | |||
XM_005268502.3 | 20 | Intron | XP_005268559.1 | |||
XM_005268503.3 | 20 | Intron | XP_005268560.1 | |||
XM_005268504.3 | 20 | Intron | XP_005268561.1 | |||
XM_005268505.3 | 20 | Intron | XP_005268562.1 | |||
XM_005268506.3 | 20 | Intron | XP_005268563.1 | |||
XM_005268507.3 | 20 | Intron | XP_005268564.1 | |||
XM_005268508.3 | 20 | Intron | XP_005268565.1 | |||
XM_005268509.3 | 20 | Intron | XP_005268566.1 | |||
XM_011537678.2 | 20 | Intron | XP_011535980.1 | |||
XM_017009792.1 | 20 | Intron | XP_016865281.1 | |||
XM_017009793.1 | 20 | Intron | XP_016865282.1 | |||
XM_017009794.1 | 20 | Intron | XP_016865283.1 | |||
XM_017009795.1 | 20 | Intron | XP_016865284.1 |