Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611241 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPRIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPRIN3 - GPRIN family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198281.2 | 7962 | Intron | NP_938022.2 | |||
XM_005262936.3 | 7962 | UTR 3 | XP_005262993.1 | |||
XM_005262937.4 | 7962 | UTR 3 | XP_005262994.1 | |||
XM_017008043.1 | 7962 | UTR 3 | XP_016863532.1 | |||
XM_017008044.1 | 7962 | UTR 3 | XP_016863533.1 |