Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600864 MIM: 603877 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CSNK1D - casein kinase 1 delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001893.4 | 2329 | Intron | NP_001884.2 | |||
NM_139062.2 | 2329 | Intron | NP_620693.1 | |||
XM_005256336.3 | 2329 | UTR 3 | XP_005256393.1 | |||
XM_005256337.4 | 2329 | Intron | XP_005256394.1 | |||
XM_017024199.1 | 2329 | UTR 3 | XP_016879688.1 |
SLC16A3 - solute carrier family 16 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |