Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600691 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC27A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC27A1 - solute carrier family 27 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198580.2 | Intron | NP_940982.1 | ||||
XM_011528000.1 | Intron | XP_011526302.1 | ||||
XM_011528001.2 | Intron | XP_011526303.1 | ||||
XM_011528002.2 | Intron | XP_011526304.1 | ||||
XM_011528003.2 | Intron | XP_011526305.1 | ||||
XM_017026781.1 | Intron | XP_016882270.1 |
Set Membership: |
HapMap |