Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE4 - copine 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289112.1 | Intron | NP_001276041.1 | ||||
NM_130808.2 | Intron | NP_570720.1 | ||||
NM_153429.1 | Intron | NP_702907.1 | ||||
XM_011512406.2 | Intron | XP_011510708.1 | ||||
XM_011512407.2 | Intron | XP_011510709.1 | ||||
XM_011512408.2 | Intron | XP_011510710.1 | ||||
XM_017005693.1 | Intron | XP_016861182.1 | ||||
XM_017005694.1 | Intron | XP_016861183.1 | ||||
XM_017005695.1 | Intron | XP_016861184.1 | ||||
XM_017005696.1 | Intron | XP_016861185.1 |