Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTCAGAAGCCTGTAGGGACCA[A/G]ACAGGTGCCCAAGATAATAATAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614364 | ||||||||||||||||||||
Literature Links: |
AACS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AACS - acetoacetyl-CoA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319839.1 | Intron | NP_001306768.1 | ||||
NM_001319840.1 | Intron | NP_001306769.1 | ||||
NM_023928.4 | Intron | NP_076417.2 |