Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BCL2L13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL2L13 - BCL2 like 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270726.1 | 263 | Silent Mutation | CCC,CCG | P,P 51 | NP_001257655.1 | |
NM_001270727.1 | 263 | Silent Mutation | CCC,CCG | P,P 51 | NP_001257656.1 | |
NM_001270728.1 | 263 | UTR 5 | NP_001257657.1 | |||
NM_001270729.1 | 263 | UTR 5 | NP_001257658.1 | |||
NM_001270730.1 | 263 | UTR 5 | NP_001257659.1 | |||
NM_001270731.1 | 263 | UTR 5 | NP_001257660.1 | |||
NM_001270732.1 | 263 | UTR 5 | NP_001257661.1 | |||
NM_001270733.1 | 263 | Intron | NP_001257662.1 | |||
NM_001270734.1 | 263 | UTR 5 | NP_001257663.1 | |||
NM_001270735.1 | 263 | UTR 5 | NP_001257664.1 | |||
NM_015367.3 | 263 | UTR 5 | NP_056182.2 |