Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACAATGGCTAACAAGAACAGGA[C/T]GACAGTCAAGTCAGTAGCTTTTTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603424 | |||||||||||||||||||||||
Literature Links: |
LOC101927027 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LOC101927027 - uncharacterized LOC101927027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKRA - protein activator of interferon induced protein kinase EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139517.1 | 1737 | UTR 3 | NP_001132989.1 | |||
NM_001139518.1 | 1737 | UTR 3 | NP_001132990.1 | |||
NM_001316362.1 | 1737 | UTR 3 | NP_001303291.1 | |||
NM_003690.4 | 1737 | UTR 3 | NP_003681.1 | |||
XM_011512063.1 | 1737 | UTR 3 | XP_011510365.1 | |||
XM_011512066.1 | 1737 | UTR 3 | XP_011510368.1 | |||
XM_017005159.1 | 1737 | UTR 3 | XP_016860648.1 |
Set Membership: |
HapMap |