Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC35C2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC35C2 - solute carrier family 35 member C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281457.1 | Intron | NP_001268386.1 | ||||
NM_001281458.1 | Intron | NP_001268387.1 | ||||
NM_001281459.1 | Intron | NP_001268388.1 | ||||
NM_001281460.1 | Intron | NP_001268389.1 | ||||
NM_015945.11 | Intron | NP_057029.8 | ||||
NM_173073.3 | Intron | NP_775096.1 | ||||
NM_173179.3 | Intron | NP_775271.1 |