Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616238 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYAR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LYAR - Ly1 antibody reactive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB49 - zinc finger and BTB domain containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145291.3 | Intron | NP_660334.3 | ||||
XM_005247951.4 | Intron | XP_005248008.1 | ||||
XM_006713864.3 | Intron | XP_006713927.1 | ||||
XM_011513416.1 | Intron | XP_011511718.1 | ||||
XM_017007835.1 | Intron | XP_016863324.1 | ||||
XM_017007836.1 | Intron | XP_016863325.1 |
Set Membership: |
HapMap |