Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138090 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
H6PD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
H6PD - hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282587.1 | 348 | Intron | NP_001269516.1 | |||
NM_004285.3 | 348 | Intron | NP_004276.2 | |||
XM_005263540.4 | 348 | Intron | XP_005263597.1 | |||
XM_006711052.3 | 348 | Intron | XP_006711115.1 | |||
XM_017002865.1 | 348 | UTR 5 | XP_016858354.1 | |||
XM_017002866.1 | 348 | Intron | XP_016858355.1 |
Set Membership: |
HapMap |