Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VPS8 - VPS8, CORVET complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009921.2 | Intron | NP_001009921.1 | ||||
NM_015303.3 | Intron | NP_056118.2 | ||||
XM_005247251.3 | Intron | XP_005247308.1 | ||||
XM_005247253.4 | Intron | XP_005247310.1 | ||||
XM_006713556.3 | Intron | XP_006713619.1 | ||||
XM_011512599.2 | Intron | XP_011510901.1 | ||||
XM_011512600.2 | Intron | XP_011510902.1 | ||||
XM_011512601.2 | Intron | XP_011510903.1 | ||||
XM_017006040.1 | Intron | XP_016861529.1 | ||||
XM_017006041.1 | Intron | XP_016861530.1 |
Set Membership: |
HapMap |