Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609387 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMC6 - structural maintenance of chromosomes 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142286.1 | Intron | NP_001135758.1 | ||||
NM_024624.5 | Intron | NP_078900.1 | ||||
XM_011533107.2 | Intron | XP_011531409.1 | ||||
XM_011533108.2 | Intron | XP_011531410.1 | ||||
XM_017004913.1 | Intron | XP_016860402.1 | ||||
XM_017004914.1 | Intron | XP_016860403.1 | ||||
XM_017004915.1 | Intron | XP_016860404.1 | ||||
XM_017004916.1 | Intron | XP_016860405.1 |