Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613726 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANO10 - anoctamin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204831.1 | Intron | NP_001191760.1 | ||||
NM_001204832.1 | Intron | NP_001191761.1 | ||||
NM_001204833.1 | Intron | NP_001191762.1 | ||||
NM_001204834.1 | Intron | NP_001191763.1 | ||||
NM_018075.3 | Intron | NP_060545.3 | ||||
XM_011533885.2 | Intron | XP_011532187.2 | ||||
XM_011533887.2 | Intron | XP_011532189.1 | ||||
XM_011533889.2 | Intron | XP_011532191.1 | ||||
XM_011533890.2 | Intron | XP_011532192.1 | ||||
XM_017006716.1 | Intron | XP_016862205.1 | ||||
XM_017006717.1 | Intron | XP_016862206.1 | ||||
XM_017006718.1 | Intron | XP_016862207.1 | ||||
XM_017006719.1 | Intron | XP_016862208.1 | ||||
XM_017006720.1 | Intron | XP_016862209.1 | ||||
XM_017006721.1 | Intron | XP_016862210.1 | ||||
XM_017006722.1 | Intron | XP_016862211.1 |