Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGGACCTGTGTGGCTTCCACG[A/G]TGGTTTTATATGTGGTTCCCCTTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604755 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372476 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105372476 - uncharacterized LOC105372476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF460 - zinc finger protein 460 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006635.3 | Intron | NP_006626.3 | ||||
XM_005258422.3 | Intron | XP_005258479.1 | ||||
XM_017026176.1 | Intron | XP_016881665.1 |
Set Membership: |
HapMap |