Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601387 MIM: 610985 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSG101 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TSG101 - tumor susceptibility 101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UEVLD - UEV and lactate/malate dehyrogenase domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040697.2 | 1577 | UTR 3 | NP_001035787.1 | |||
NM_001261382.1 | 1577 | UTR 3 | NP_001248311.1 | |||
NM_001261383.1 | 1577 | UTR 3 | NP_001248312.1 | |||
NM_001261384.1 | 1577 | UTR 3 | NP_001248313.1 | |||
NM_001261385.1 | 1577 | UTR 3 | NP_001248314.1 | |||
NM_001261386.1 | 1577 | UTR 3 | NP_001248315.1 | |||
NM_001297771.1 | 1577 | Intron | NP_001284700.1 | |||
NM_018314.4 | 1577 | UTR 3 | NP_060784.3 | |||
XM_006718255.2 | 1577 | UTR 3 | XP_006718318.1 | |||
XM_011520201.2 | 1577 | UTR 3 | XP_011518503.1 | |||
XM_017017985.1 | 1577 | UTR 3 | XP_016873474.1 | |||
XM_017017986.1 | 1577 | UTR 3 | XP_016873475.1 |
Set Membership: |
HapMap |