Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP170B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP170B - centrosomal protein 170B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001112726.2 | Intron | NP_001106197.1 | ||||
NM_015005.2 | Intron | NP_055820.2 | ||||
XM_005267550.3 | Intron | XP_005267607.1 | ||||
XM_011536665.2 | Intron | XP_011534967.1 | ||||
XM_011536666.2 | Intron | XP_011534968.1 | ||||
XM_011536667.2 | Intron | XP_011534969.1 | ||||
XM_017021227.1 | Intron | XP_016876716.1 | ||||
XM_017021228.1 | Intron | XP_016876717.1 | ||||
XM_017021229.1 | Intron | XP_016876718.1 | ||||
XM_017021230.1 | Intron | XP_016876719.1 |
Set Membership: |
HapMap |