Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601207 MIM: 616660 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKQ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGKQ - diacylglycerol kinase theta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001347.3 | 3326 | UTR 3 | NP_001338.2 | |||
XM_011513411.1 | 3326 | UTR 3 | XP_011511713.1 | |||
XM_011513412.1 | 3326 | Intron | XP_011511714.1 | |||
XM_011513414.2 | 3326 | Intron | XP_011511716.1 | |||
XM_011513415.1 | 3326 | Intron | XP_011511717.1 | |||
XM_017007814.1 | 3326 | Intron | XP_016863303.1 | |||
XM_017007815.1 | 3326 | Intron | XP_016863304.1 |
TMEM175 - transmembrane protein 175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |