Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607990 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRDC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARRDC5 - arrestin domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080523.1 | Intron | NP_001073992.1 | ||||
XM_005259616.2 | Intron | XP_005259673.1 | ||||
XM_011528181.2 | Intron | XP_011526483.1 | ||||
XM_017027144.1 | Intron | XP_016882633.1 | ||||
XM_017027145.1 | Intron | XP_016882634.1 |
UHRF1 - ubiquitin like with PHD and ring finger domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048201.2 | Intron | NP_001041666.1 | ||||
NM_001290050.1 | Intron | NP_001276979.1 | ||||
NM_001290051.1 | Intron | NP_001276980.1 | ||||
NM_001290052.1 | Intron | NP_001276981.1 | ||||
NM_013282.4 | Intron | NP_037414.3 | ||||
XM_011527942.2 | Intron | XP_011526244.1 |
Set Membership: |
HapMap |