Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf42 - chromosome 11 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173525.2 | 590 | Intron | NP_775796.2 | |||
XM_011519926.2 | 590 | UTR 5 | XP_011518228.1 | |||
XM_011519927.2 | 590 | UTR 5 | XP_011518229.1 |
FAM160A2 - family with sequence similarity 160 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR52W1 - olfactory receptor family 52 subfamily W member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |