Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604621 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGAT3 - mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098270.1 | Intron | NP_001091740.1 | ||||
NM_002409.4 | Intron | NP_002400.3 | ||||
XM_005261608.4 | Intron | XP_005261665.1 | ||||
XM_005261609.2 | Intron | XP_005261666.1 | ||||
XM_006724252.2 | Intron | XP_006724315.1 | ||||
XM_011530184.2 | Intron | XP_011528486.1 | ||||
XM_011530185.2 | Intron | XP_011528487.1 | ||||
XM_011530186.2 | Intron | XP_011528488.1 | ||||
XM_011530187.2 | Intron | XP_011528489.1 | ||||
XM_011530188.2 | Intron | XP_011528490.1 | ||||
XM_011530189.2 | Intron | XP_011528491.1 | ||||
XM_011530190.2 | Intron | XP_011528492.1 | ||||
XM_011530191.2 | Intron | XP_011528493.1 | ||||
XM_011530192.2 | Intron | XP_011528494.1 | ||||
XM_011530194.2 | Intron | XP_011528496.1 | ||||
XM_017028802.1 | Intron | XP_016884291.1 |