Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACTCAGACGGGGCAGTGGGTT[C/T]TTCTGGGCTGCAAGTTTGTTATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP8 - Rho GTPase activating protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017526.1 | Intron | NP_001017526.1 | ||||
NM_001198726.1 | Intron | NP_001185655.1 | ||||
NM_181335.2 | Intron | NP_851852.2 |
PRR5-ARHGAP8 - PRR5-ARHGAP8 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181334.5 | Intron | NP_851851.3 |