Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607934 MIM: 606755 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PADI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PADI1 - peptidyl arginine deiminase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PADI3 - peptidyl arginine deiminase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016233.2 | Intron | NP_057317.2 | ||||
XM_011541571.2 | Intron | XP_011539873.1 | ||||
XM_011541572.2 | Intron | XP_011539874.1 | ||||
XM_017001463.1 | Intron | XP_016856952.1 |